siRNA Details
| Gene Symbol: Unc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153135.3 | NP_694775.1 | 10, 11 | 1618 | ||
| GenBank mRNA | AJ487854.1 | - | 10, 11 | 1619 | ||
| BC137615.1 | - | 10, 11 | 1409 | 8860634 | ||
| BC144783.1 | - | 8, 9 | 1226 | 9053331 | ||
| BC145695.1 | - | 7, 8 | 1193 | 40130998 | ||

Unc5d
Genomic Map