siRNA Details
| Gene Symbol: BC037034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153161.3 | NP_694801.2 | 11 | 2113 | ||
| GenBank mRNA | AK046882.1 | - | 7 | 3136 | ||
| AK138960.1 | - | 11 | 2057 | |||
| BC037034.1 | - | 10 | 1516 | 4505093 | ||
| BC040828.1 | - | 1 | 2086 | |||
| BC044860.1 | - | 1 | 2098 | |||

BC037034
Genomic Map