siRNA Details
| Gene Symbol: Klhl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039059.1 | NP_001034148.1 | 3 | 914 | ||
| NM_001039060.1 | NP_001034149.1 | 4 | 965 | |||
| NM_001039061.1 | NP_001034150.1 | 2 | 1009 | |||
| NM_153165.2 | NP_694805.1 | 4 | 965 | |||
| GenBank mRNA | AK078112.1 | - | 2 | 950 | ||
| AK081546.1 | - | 3 | 918 | |||
| AK133055.1 | - | 3 | 679 | |||
| AK169500.1 | - | 4 | 908 | |||
| BC036978.1 | - | 4 | 902 | |||
| BC141165.1 | - | 3 | 679 | 9055777 | ||

Klhl15
Genomic Map