siRNA Details
| Gene Symbol: Pnma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153169.2 | NP_694809.1 | 1 | 1758 | ||
| GenBank mRNA | AK141560.1 | - | 1 | 1227 | ||
| BC036726.1 | - | 1 | 1796 | 5368858 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnma3| Gene Symbol: Pnma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153169.2 | NP_694809.1 | 1 | 1758 | ||
| GenBank mRNA | AK141560.1 | - | 1 | 1227 | ||
| BC036726.1 | - | 1 | 1796 | 5368858 | ||