siRNA Details
| Gene Symbol: Fbxo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153195.2 | NP_694875.2 | 3 | 523 | ||
| GenBank mRNA | AK147797.1 | - | 3 | 475 | ||
| AK147907.1 | - | 3 | 442 | |||
| AK150190.1 | - | 3 | 298 | |||
| AK150626.1 | - | 3 | 481 | |||
| AK151025.1 | - | 3 | 481 | |||
| AK152513.1 | - | 3 | 481 | |||
| AK182909.1 | - | 2 | 135 | |||
| BC032153.1 | - | 3 | 465 | 5054087 | ||

Fbxo7
Genomic Map