siRNA Details
| Gene Symbol: P2rx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164833.1 | NP_001158305.1 | 1 | 187 | ||
| NM_001164834.1 | NP_001158306.1 | 1 | 187 | |||
| NM_153400.4 | NP_700449.2 | 1 | 187 | |||
| GenBank mRNA | AB094663.1 | - | 1 | 107 | ||
| AB094664.1 | - | 1 | 107 | |||
| AK141196.1 | - | 1 | 146 | |||
| AK158334.1 | - | 1 | 129 | |||
| AY044240.2 | - | 1 | 363 | |||
| BY236715.1 | - | 1 | 190 | |||

P2rx2
Genomic Map