siRNA Details
| Gene Symbol: Dock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153413.2 | NP_700462.2 | 2 | 479 | ||
| GenBank mRNA | AK172930.1 | - | 2 | 438 | ||
| AY145302.1 | - | 2 | 429 | |||
| BF465833.1 | - | 2 | 153 | |||
| CJ124323.1 | - | 2 | 479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dock3| Gene Symbol: Dock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153413.2 | NP_700462.2 | 2 | 479 | ||
| GenBank mRNA | AK172930.1 | - | 2 | 438 | ||
| AY145302.1 | - | 2 | 429 | |||
| BF465833.1 | - | 2 | 153 | |||
| CJ124323.1 | - | 2 | 479 | |||