siRNA Details
| Gene Symbol: Dusp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153459.4 | NP_703189.3 | 1, 2 | 700 | ||
| GenBank mRNA | AK140372.1 | - | 1, 2 | 701 | ||
| BC010207.1 | - | 1, 2 | 343 | 3964527 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp7| Gene Symbol: Dusp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153459.4 | NP_703189.3 | 1, 2 | 700 | ||
| GenBank mRNA | AK140372.1 | - | 1, 2 | 701 | ||
| BC010207.1 | - | 1, 2 | 343 | 3964527 | ||