siRNA Details
Gene Symbol: Clec2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153506.4 | NP_705726.3 | 5 | 561 | ||
GenBank mRNA | BC051091.1 | - | 5 | 575 | 6529908 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Clec2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153506.4 | NP_705726.3 | 5 | 561 | ||
GenBank mRNA | BC051091.1 | - | 5 | 575 | 6529908 |