siRNA Details
| Gene Symbol: Nrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153529.2 | NP_705757.1 | 2 | 303 | ||
| GenBank mRNA | AK003046.1 | - | 2 | 130 | ||
| AK161859.1 | - | 2 | 271 | |||
| BC035531.1 | - | 2 | 291 | 5367281 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrn1| Gene Symbol: Nrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153529.2 | NP_705757.1 | 2 | 303 | ||
| GenBank mRNA | AK003046.1 | - | 2 | 130 | ||
| AK161859.1 | - | 2 | 271 | |||
| BC035531.1 | - | 2 | 291 | 5367281 | ||