siRNA Details
| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153535.2 | NP_705763.1 | 1 | 805 | ||
| GenBank mRNA | AK140440.1 | - | 1 | 805 | ||
| AK141607.1 | - | 1 | 748 | |||
| BC035537.1 | - | 1 | 696 | 5369152 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam149a| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153535.2 | NP_705763.1 | 1 | 805 | ||
| GenBank mRNA | AK140440.1 | - | 1 | 805 | ||
| AK141607.1 | - | 1 | 748 | |||
| BC035537.1 | - | 1 | 696 | 5369152 | ||