siRNA Details
| Gene Symbol: Evi5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039578.3 | NP_001034667.3 | 3 | 559 | ||
| GenBank mRNA | AK045240.1 | - | 3 | 560 | ||
| AK087482.1 | - | 3 | 560 | |||
| BC042653.1 | - | 3 | 539 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Evi5l| Gene Symbol: Evi5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039578.3 | NP_001034667.3 | 3 | 559 | ||
| GenBank mRNA | AK045240.1 | - | 3 | 560 | ||
| AK087482.1 | - | 3 | 560 | |||
| BC042653.1 | - | 3 | 539 | |||