siRNA Details
| Gene Symbol: Dusp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153459.4 | NP_703189.3 | 2 | 957 | ||
| GenBank mRNA | AK140372.1 | - | 2 | 958 | ||
| AK158817.1 | - | 1 | 133 | |||
| BC010207.1 | - | 2 | 600 | 3964527 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp7| Gene Symbol: Dusp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153459.4 | NP_703189.3 | 2 | 957 | ||
| GenBank mRNA | AK140372.1 | - | 2 | 958 | ||
| AK158817.1 | - | 1 | 133 | |||
| BC010207.1 | - | 2 | 600 | 3964527 | ||