siRNA Details
Gene Symbol: Rnf183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153504.3 | NP_705724.1 | 3 | 989 | ||
GenBank mRNA | AK017982.1 | - | 3 | 990 | ||
BC025512.1 | - | 2 | 751 | 5252940 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnf183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153504.3 | NP_705724.1 | 3 | 989 | ||
GenBank mRNA | AK017982.1 | - | 3 | 990 | ||
BC025512.1 | - | 2 | 751 | 5252940 |