siRNA Details
| Gene Symbol: Cpne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153507.2 | NP_705727.1 | 4, 5 | 635 | ||
| GenBank mRNA | BC022128.1 | - | 4, 5 | 596 | 5097527 | |
| BC031801.1 | - | 4, 5 | 635 | 3985959 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpne2| Gene Symbol: Cpne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153507.2 | NP_705727.1 | 4, 5 | 635 | ||
| GenBank mRNA | BC022128.1 | - | 4, 5 | 596 | 5097527 | |
| BC031801.1 | - | 4, 5 | 635 | 3985959 | ||