siRNA Details
Gene Symbol: BC028528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153513.2 | NP_705733.2 | 3, 4 | 191 | ||
GenBank mRNA | AF534879.1 | - | 3, 4 | 191 | ||
BC028528.1 | - | 3, 4 | 209 | 1366537 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC028528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153513.2 | NP_705733.2 | 3, 4 | 191 | ||
GenBank mRNA | AF534879.1 | - | 3, 4 | 191 | ||
BC028528.1 | - | 3, 4 | 209 | 1366537 |