siRNA Details
| Gene Symbol: Ccdc65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153518.1 | NP_705738.1 | 1, 2 | 318 | ||
| GenBank mRNA | AF454432.1 | - | 1, 2 | 318 | ||
| AK077174.1 | - | 1, 2 | 352 | |||
| BC048073.1 | - | 1, 2 | 338 | 6312435 | ||
| BC056947.1 | - | 1, 2 | 314 | |||
| BC060155.1 | - | 1, 2 | 314 | |||

Ccdc65
Genomic Map