siRNA Details
| Gene Symbol: Zfp280c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166648.1 | NP_001160120.1 | 7, 8 | 663 | ||
| NM_001166649.1 | NP_001160121.1 | 7, 8 | 684 | |||
| NM_001166650.1 | NP_001160122.1 | 6, 7 | 622 | |||
| NM_153532.3 | NP_705760.2 | 6, 7 | 994 | |||
| GenBank mRNA | AK129399.1 | - | 7, 8 | 643 | ||
| AK141491.1 | - | 7, 8 | 664 | |||
| BC028839.1 | - | 6, 7 | 634 | 5150255 | ||
| BC051397.1 | - | 6, 7 | 622 | 5150255 | ||
| BC063775.1 | - | 6, 7 | 1006 | 5375514 | ||

Zfp280c
Genomic Map