siRNA Details
| Gene Symbol: BC030867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153544.3 | NP_705772.3 | 3 | 564 | ||
| GenBank mRNA | AK029811.1 | - | 3 | 562 | ||
| BC030867.1 | - | 3 | 547 | |||
| BC085317.1 | - | 3 | 483 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC030867| Gene Symbol: BC030867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153544.3 | NP_705772.3 | 3 | 564 | ||
| GenBank mRNA | AK029811.1 | - | 3 | 562 | ||
| BC030867.1 | - | 3 | 547 | |||
| BC085317.1 | - | 3 | 483 | |||