siRNA Details
| Gene Symbol: Gnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153547.6 | NP_705775.2 | 3 | 296 | ||
| GenBank mRNA | AK077523.1 | - | 3 | 227 | ||
| AK087757.1 | - | 3 | 180 | |||
| AK167094.1 | - | 1 | 1715 | |||
| AK167183.1 | - | 3 | 180 | |||
| AY181025.1 | - | 3 | 154 | |||
| AY185498.1 | - | 3 | 154 | |||
| BC018560.1 | - | 3 | 183 | |||
| BC037996.1 | - | 3 | 156 | 5052858 | ||
| BY332148.1 | - | 3 | 255 | |||
| CJ071960.1 | - | 3 | 299 | |||

Gnl3
Genomic Map