siRNA Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153548.2 | NP_705776.2 | 12, 13 | 1761 | ||
| GenBank mRNA | AK128970.1 | - | 12, 13 | 1776 | ||
| AK205606.1 | - | 2, 3 | 165 | |||
| BC034070.1 | - | 2, 3 | 361 | 4217923 | ||
| BC040815.1 | - | 2, 3 | 361 | 4217923 | ||
| BC067035.1 | - | 12, 13 | 1761 | 30532658 | ||

E430025E21Rik
Genomic Map