siRNA Details
| Gene Symbol: Fam102a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153560.4 | NP_705788.1 | 2 | 895 | ||
| GenBank mRNA | AK049032.1 | - | 2 | 899 | ||
| AK131145.1 | - | 2 | 922 | |||
| AK157331.1 | - | 2 | 525 | |||
| AK157490.1 | - | 3 | 317 | |||
| AK158435.1 | - | 2 | 522 | |||
| AK165751.1 | - | 2 | 525 | |||
| AK170536.1 | - | 2 | 923 | |||
| AK170847.1 | - | 2 | 516 | |||
| AK170951.1 | - | 2 | 522 | |||
| AK172536.1 | - | 2 | 522 | |||
| AK205749.1 | - | 2 | 48 | |||
| BC023752.1 | - | 2 | 340 | 5354455 | ||
| BC031157.1 | - | 2 | 158 | 4976773 | ||

Fam102a
Genomic Map