siRNA Details
| Gene Symbol: Dock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153413.2 | NP_700462.2 | 32 | 3789 | ||
| GenBank mRNA | AK147385.1 | - | 18 | 2102 | ||
| AY145302.1 | - | 32 | 3730 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dock3| Gene Symbol: Dock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153413.2 | NP_700462.2 | 32 | 3789 | ||
| GenBank mRNA | AK147385.1 | - | 18 | 2102 | ||
| AY145302.1 | - | 32 | 3730 | |||