siRNA Details
| Gene Symbol: Rhobtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153514.5 | NP_705734.4 | 10 | 3858 | ||
| GenBank mRNA | AK081100.1 | - | 10 | 3787 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhobtb2| Gene Symbol: Rhobtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153514.5 | NP_705734.4 | 10 | 3858 | ||
| GenBank mRNA | AK081100.1 | - | 10 | 3787 | ||