siRNA Details
| Gene Symbol: Gramd1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172107.1 | NP_001165578.1 | 5 | 566 | ||
| NM_153528.2 | NP_705756.1 | 5 | 796 | |||
| GenBank mRNA | AK029543.1 | - | 5 | 567 | ||
| AK035257.1 | - | 5 | 799 | |||
| AK129016.1 | - | 5 | 735 | |||
| BC037472.1 | - | 5 | 730 | 4236901 | ||

Gramd1c
Genomic Map