siRNA Details
Gene Symbol: Zfp280c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166648.1 | NP_001160120.1 | 15 | 1974 | ||
NM_001166649.1 | NP_001160121.1 | 15 | 1995 | |||
NM_001166650.1 | NP_001160122.1 | 14 | 1933 | |||
NM_153532.3 | NP_705760.2 | 14 | 2305 | |||
GenBank mRNA | AK129399.1 | - | 15 | 1954 | ||
AK141491.1 | - | 15 | 1975 | |||
BC028839.1 | - | 14 | 1945 | 5150255 | ||
BC051397.1 | - | 14 | 1933 | 5150255 | ||
BC063775.1 | - | 14 | 2317 | 5375514 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
