siRNA Details
| Gene Symbol: Evi5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006992.1 | - | 3 | 1705 | ||
| AK155998.1 | - | 7 | 2427 | |||
| BC042653.1 | - | 18 | 4029 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Evi5l| Gene Symbol: Evi5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006992.1 | - | 3 | 1705 | ||
| AK155998.1 | - | 7 | 2427 | |||
| BC042653.1 | - | 18 | 4029 | |||