siRNA Details
| Gene Symbol: Lrrc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153542.1 | NP_705770.1 | 5 | 975 | ||
| GenBank mRNA | AK136380.1 | - | 5 | 1002 | ||
| AK139384.1 | - | 5 | 1008 | |||
| AK145920.1 | - | 5 | 992 | |||
| AK150142.1 | - | 5 | 976 | |||
| AK150246.1 | - | 5 | 976 | |||
| BC036304.1 | - | 5 | 975 | 3968534 | ||

Lrrc20
Genomic Map