siRNA Details
| Gene Symbol: BC030867 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153544.3 | NP_705772.3 | 3 | 636 | ||
| GenBank mRNA | AK029811.1 | - | 3 | 634 | ||
| BC030867.1 | - | 3 | 619 | |||
| BC085317.1 | - | 3 | 555 | |||
| BC108338.1 | - | 3 | 617 | 5034745 | ||

BC030867
Genomic Map