siRNA Details
| Gene Symbol: UGT2B15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076.3 | NP_001067.2 | 6 | 1981 | ||
| GenBank mRNA | AF180322.1 | - | 6 | 1862 | ||
| AF548389.1 | - | 6 | 1841 | |||
| BC171879.1 | - | 2759 | 9054533 | |||
| U08854.1 | - | 6 | 1845 | |||

UGT2B15
Genomic Map