siRNA Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153555.2 | NP_705783.1 | 10 | 1524 | ||
| GenBank mRNA | AK035701.1 | - | 10 | 1423 | ||
| AK098114.1 | - | 10 | 1543 | |||
| AK146825.1 | - | 10 | 1451 | |||
| AK162622.1 | - | 8 | 2472 | |||
| AK164799.1 | - | 10 | 1451 | |||
| AK167059.1 | - | 10 | 1400 | |||
| BC023704.1 | - | 10 | 1407 | 5345701 | ||
| BC023804.1 | - | 11 | 1578 | 5352941 | ||
| BC078641.1 | - | 10 | 2436 | |||

Dcaf8
Genomic Map