siRNA Details
Gene Symbol: BC029214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153557.1 | NP_705785.1 | 7 | 810 | ||
GenBank mRNA | AK053101.1 | - | 5 | 422 | ||
BC029214.1 | - | 7 | 810 | 4161391 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC029214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153557.1 | NP_705785.1 | 7 | 810 | ||
GenBank mRNA | AK053101.1 | - | 5 | 422 | ||
BC029214.1 | - | 7 | 810 | 4161391 |