siRNA Details
| Gene Symbol: Fam13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153574.2 | NP_705802.1 | 4 | 304 | ||
| GenBank mRNA | AJ510136.1 | - | 4 | 290 | ||
| AK052421.1 | - | 4 | 245 | |||
| AK054311.1 | - | 4 | 291 | |||
| BC005554.1 | - | 4 | 209 | |||
| BC054053.1 | - | 4 | 278 | |||
| BC060978.1 | - | 4 | 241 | |||
| BC080861.1 | - | 4 | 304 | 30357292 | ||

Fam13a
Genomic Map