siRNA Details
| Gene Symbol: Cxcl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153576.2 | NP_705804.2 | 2 | 250 | ||
| GenBank mRNA | AK143005.1 | - | 2 | 166 | ||
| AY374329.1 | - | 2 | 153 | |||
| AY598463.1 | - | 2 | 251 | |||
| BC024561.1 | - | 2 | 250 | 5004999 | ||

Cxcl17
Genomic Map