siRNA Details
| Gene Symbol: Sv2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109753.1 | NP_001103223.1 | 2 | 653 | ||
| NM_153579.4 | NP_705807.2 | 2 | 860 | |||
| GenBank mRNA | AK029736.1 | - | 3 | 685 | ||
| AK035927.1 | - | 2 | 578 | |||
| AK043830.1 | - | 2 | 580 | |||
| AK139478.1 | - | 2 | 555 | |||
| AK140104.1 | - | 2 | 573 | |||
| AK147261.1 | - | 2 | 647 | |||
| AK159028.1 | - | 2 | 558 | |||
| BC060224.1 | - | 1 | 244 | 30356990 | ||
| BC079568.1 | - | 2 | 860 | |||

Sv2b
Genomic Map