siRNA Details
Gene Symbol: Trpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153597.2 | NP_705825.2 | 4 | 772 | ||
GenBank mRNA | AK038830.1 | - | 3 | 604 | ||
AK048558.1 | - | 921 | ||||
AK090183.1 | - | 3 | 1903 | |||
AK141582.1 | - | 559 | ||||
BC027575.1 | - | 4 | 570 | 1364225 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
