siRNA Details
| Gene Symbol: Snx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153680.2 | NP_710147.1 | 3 | 331 | ||
| GenBank mRNA | AK047265.1 | - | 3 | 331 | ||
| AK077650.1 | - | 3 | 333 | |||
| AK077780.1 | - | 3 | 333 | |||
| AK156299.1 | - | 3 | 382 | |||
| AK190219.1 | - | 1 | 71 | |||
| AK207583.1 | - | 1 | 71 | |||
| BC023732.1 | - | 3 | 347 | 5352355 | ||
| BC026571.1 | - | 3 | 331 | 4986268 | ||
| BC029675.1 | - | 3 | 318 | |||

Snx17
Genomic Map