siRNA Details
| Gene Symbol: Rnf26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153762.3 | NP_717095.2 | 1 | 288 | ||
| GenBank mRNA | AK033164.1 | - | 1 | 742 | ||
| AK085078.1 | - | 738 | ||||
| AK157821.1 | - | 1 | 737 | |||
| BC004739.1 | - | 1 | 339 | 3501511 | ||
| BC063251.1 | - | 1 | 288 | 6413980 | ||
| BC086757.1 | - | 1 | 271 | |||

Rnf26
Genomic Map