siRNA Details
| Gene Symbol: Pygb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153781.1 | NP_722476.1 | 3 | 452 | ||
| GenBank mRNA | AK032769.1 | - | 3 | 465 | ||
| AK035673.1 | - | 3 | 181 | |||
| AK134667.1 | - | 3 | 462 | |||
| AK142716.1 | - | 3 | 461 | |||
| AK147763.1 | - | 3 | 484 | |||
| AK168381.1 | - | 3 | 462 | |||
| AK169052.1 | - | 3 | 487 | |||
| AK169631.1 | - | 3 | 466 | |||
| BC031457.1 | - | 3 | 460 | |||
| BC035283.1 | - | 3 | 452 | 4947455 | ||

Pygb
Genomic Map