siRNA Details
| Gene Symbol: Zfp128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153802.4 | NP_722497.1 | 2 | 265 | ||
| GenBank mRNA | AF480861.1 | - | 2 | 223 | ||
| AK032046.1 | - | 2 | 245 | |||
| AK035906.1 | - | 2 | 266 | |||
| AK132244.1 | - | 2 | 267 | |||
| BC088998.1 | - | 2 | 236 | 6417536 | ||
| BC094248.1 | - | 2 | 236 | 5718665 | ||

Zfp128
Genomic Map