siRNA Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170591.1 | NP_733479.1 | 2 | 313 | ||
| GenBank mRNA | AK006062.1 | - | 2 | 415 | ||
| AK029688.1 | - | 367 | ||||
| AK078279.1 | - | 2 | 361 | |||
| AK086412.1 | - | 2 | 381 | |||
| AK129134.1 | - | 2 | 350 | |||
| AK133721.1 | - | 2 | 324 | |||
| AK139895.1 | - | 2 | 443 | |||
| AK146477.1 | - | 2 | 313 | |||
| AK163792.1 | - | 2 | 342 | |||
| AK194884.1 | - | 2 | 127 | |||
| BC026743.1 | - | 2 | 344 | 3964978 | ||

Nupl1
Genomic Map