siRNA Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170755.2 | NP_739561.2 | 4, 5 | 609 | ||
| GenBank mRNA | AK034667.1 | - | 4, 5 | 610 | ||
| AK152268.1 | - | 4, 5 | 324 | |||
| AK158940.1 | - | 4, 5 | 379 | |||
| AK160400.1 | - | 4, 5 | 563 | |||
| AK165957.1 | - | 4, 5 | 588 | |||
| AK172093.1 | - | 4, 5 | 606 | |||
| AK202961.1 | - | 1, 2 | 102 | |||
| BC070406.1 | - | 4, 5 | 601 | 5707319 | ||
| BY261213.1 | - | 4, 5 | 387 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001100760, XM_006245244, BC104673.

Fam134a
Genomic Map