siRNA Details
| Gene Symbol: Ugt2b34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153598.2 | NP_705826.1 | 1 | 226 | ||
| GenBank mRNA | AK145951.1 | - | 1 | 236 | ||
| BC028826.1 | - | 1 | 226 | 5098890 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt2b34| Gene Symbol: Ugt2b34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153598.2 | NP_705826.1 | 1 | 226 | ||
| GenBank mRNA | AK145951.1 | - | 1 | 236 | ||
| BC028826.1 | - | 1 | 226 | 5098890 | ||