siRNA Details
| Gene Symbol: Lrrc56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172064.1 | NP_001165535.1 | 6, 7 | 1001 | ||
| NM_001172065.1 | NP_001165536.1 | 6, 7 | 1001 | |||
| NM_153777.2 | NP_722472.1 | 6, 7 | 587 | |||
| GenBank mRNA | AK017603.1 | - | 6, 7 | 975 | ||
| AK220237.1 | - | 6, 7 | 955 | |||
| BC027807.1 | - | 6, 7 | 597 | 5324022 | ||

Lrrc56
Genomic Map