siRNA Details
| Gene Symbol: Atoh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153778.3 | NP_722473.1 | 3 | 1538 | ||
| GenBank mRNA | AB046527.1 | - | 3 | 1540 | ||
| AB046528.1 | - | 3 | 1236 | |||
| AB046529.1 | - | 3 | 498 | |||
| AB046530.1 | - | 4 | 650 | |||
| AK016909.1 | - | 3 | 489 | |||
| AY349615.1 | - | 4 | 1246 | |||
| BC023684.1 | - | 3 | 1206 | 5343472 | ||

Atoh8
Genomic Map