siRNA Details
| Gene Symbol: Gm1976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049147.1 | - | 1 | 431 | ||
| Gene Symbol: 2610044O15Rik8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153780.3 | NP_722475.1 | 5 | 608 | ||
| GenBank mRNA | AK011776.1 | - | 5 | 585 | ||
| AK029464.1 | - | 5 | 571 | |||
| AK138138.1 | - | 5 | 609 | |||
| BC023890.1 | - | 5 | 567 | 5252244 | ||

Gm1976
Genomic Map