siRNA Details
| Gene Symbol: Paox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153783.4 | NP_722478.2 | 2 | 508 | ||
| GenBank mRNA | AF226656.2 | - | 2 | 527 | ||
| AK010469.1 | - | 1 | 157 | |||
| AK030664.1 | - | 2 | 509 | |||
| AM049402.1 | - | 2 | 500 | |||
| BC082783.1 | - | 2 | 507 | 6475809 | ||

Paox
Genomic Map