siRNA Details
| Gene Symbol: Traf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172113.1 | NP_001165584.1 | 8, 9 | 876 | ||
| NM_001172114.1 | NP_001165585.1 | 7, 8 | 762 | |||
| NM_153792.2 | NP_722487.2 | 8, 9 | 896 | |||
| GenBank mRNA | AK088431.1 | - | 7, 8 | 671 | ||
| AK131617.1 | - | 8, 9 | 837 | |||
| AK178691.1 | - | 14 | ||||
| AK217262.1 | - | 14 | ||||
| BC008598.1 | - | 8, 9 | 799 | 3499933 | ||

Traf7
Genomic Map