siRNA Details
| Gene Symbol: Smc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252684.1 | NP_001239613.1 | 2 | 382 | ||
| NM_001252685.1 | NP_001239614.1 | 2 | 211 | |||
| NM_153808.2 | NP_722503.1 | 2 | 382 | |||
| GenBank mRNA | AJ534941.1 | - | 2 | 303 | ||
| AK143818.1 | - | 2 | 359 | |||
| AK161254.1 | - | 2 | 212 | |||
| BC038345.1 | - | 2 | 362 | 3596405 | ||

Smc5
Genomic Map