siRNA Details
| Gene Symbol: Igsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170599.2 | NP_733548.2 | 3 | 402 | ||
| GenBank mRNA | AB079880.1 | - | 3 | 239 | ||
| AK160146.1 | - | 3 | 365 | |||
| BC057555.1 | - | 3 | 402 | 6416190 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igsf11| Gene Symbol: Igsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170599.2 | NP_733548.2 | 3 | 402 | ||
| GenBank mRNA | AB079880.1 | - | 3 | 239 | ||
| AK160146.1 | - | 3 | 365 | |||
| BC057555.1 | - | 3 | 402 | 6416190 | ||